Documenti fondamentali
BAF133740000
SP Bel-Art® Belpen Marker Pen
black
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Materiali
black
Produttore/marchio commerciale
Bel-Art F13374-0000
Categorie correlate
Descrizione generale
Felt tip marking pens are perfect for labeling all types of glassware and plastics.
Caratteristiche e vantaggi
  • Permanent, quick drying ink remains waterproof and smudge proof
  • Will withstand autoclaving when dry
  • Pen with handy pocket clip is 13.3cm (5¹/4") long
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)