BAB609110100
SP Bel-Art® Durac® Bluetooth Thermometer with 30 day logger
temp. range at -10-50 °C
Sinonimo/i:
Verification Bluetooth Thermometer
About This Item
Codice UNSPSC:
41112303
NACRES:
NB.79
Prodotti consigliati
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
HB Instruments B60911-0100
range of application measuring range
-10-50 °C, resolution: 0.1 °F/C (±1°C (2°F) between 0/40°C (32/104°F); otherwise ±2°C (4°F))
Parametri
temp. range at -10-50 °C
Alt. × lungh. × largh.
20 mm × 60 mm × 85 mm
Descrizione generale
Enable remote monitoring and alerts by broadcasting readings to any Apple or Android smartphone or device with Bluetooth 4.0 or better utilizing the free THERMSmart app. Available in verification and environmental monitoring models.
Caratteristiche e vantaggi
- Monitor critical environments without a physical connection; maximum signal distance is approximately 40 meters
- Record temperature data for 30 days
- Hourly data recorded for current seven days, then maximum and minimum temperatures are recorded for next 23 days
- Verification model records accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample
- Alarm sounds to alert user if temperature falls outside of the set parameters
- Data viewable as a table, graph, or calendar; data from up to six data loggers can be plotted on one graph for direct comparison; also downloadable as a CSV file
- Indicators include memory full, active alarm state and low battery
- App can trigger indicator light for unit identification
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)