Skip to Content
Merck
All Photos(4)

Key Documents

V900888

Sigma-Aldrich

Thiazolyl Blue Tetrazolium Bromide

Vetec, reagent grade, 98%

Synonym(s):

3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium bromide, MTT, Methylthiazolyldiphenyl-tetrazolium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16BrN5S
CAS Number:
Molecular Weight:
414.32
Beilstein:
4081397
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

form

powder

mp

195 °C (dec.) (lit.)

solubility

H2O: 5 mg/mL, clear

suitability

suitable for

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Br-].Cc1nc(sc1C)-[n+]2nc(nn2-c3ccccc3)-c4ccccc4

InChI

1S/C18H16N5S.BrH/c1-13-14(2)24-18(19-13)23-21-17(15-9-5-3-6-10-15)20-22(23)16-11-7-4-8-12-16;/h3-12H,1-2H3;1H/q+1;/p-1

InChI key

AZKSAVLVSZKNRD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Thiazolyl Blue Tetrazolium Blue (MTT) may be used in measurement of cell proliferation. MTT produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells. The blue crystals are solubilized with acidified isopropanol and the intensity is measured colorimetrically at 570 nm. MTT has been used as a histochemical/cytochemical reagent and for the detection of NAD. ADP-linked enzyme systems in tissue cannot be detected with MTT, due to binding of the cation by the cyanide trap used. MTT is rapidly reduced to the formazan, which chelates with nickel, copper, and cobalt; the cobalt chelate has been used in oxidative systems.

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markHealth hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael G Sugiyama et al.
Scientific reports, 5, 11030-11030 (2015-06-06)
Seasonal influenza virus infections cause hundreds of thousands of deaths annually while viral mutation raises the threat of a novel pandemic strain. Antiviral drugs exhibit limited efficacy unless administered early and may induce viral resistance. Thus, targeting the host response
An-Ming Wang et al.
Oncotarget, 5(13), 5002-5016 (2014-06-28)
Gastric cancer is the second leading cause of cancer-related death worldwide. Herein, we investigated the role of transcription factor Yin Yang 1 (YY1), a multi-functional protein, in tumorigenesis of gastric cancer cells. Results showed that YY1 contributed to gastric carcinogenesis
Rute B Marques et al.
European urology, 67(6), 1177-1185 (2014-09-16)
The phosphatidylinositol-4,5-bisphosphate 3-kinase (PI3K)/AKT pathway is frequently activated during prostate cancer (PCa) progression through loss or mutation of the phosphatase and tensin homolog (PTEN) gene. Following the androgen receptor (AR) pathway, it is the second major driver of PCa growth.
V Kumar et al.
Leukemia, 28(12), 2324-2335 (2014-04-15)
Notch signaling deregulation is linked to the onset of several tumors including T-cell acute lymphoblastic leukemia (T-ALL). Deregulated microRNA (miRNA) expression is also associated with several cancers, including leukemias. However, the transcriptional regulators of miRNAs, as well as the relationships
Lisa M Kaminskas et al.
Molecular pharmaceutics, 12(2), 432-443 (2014-12-09)
The current study sought to explore whether the subcutaneous administration of lymph targeted dendrimers, conjugated with a model chemotherapeutic (methotrexate, MTX), was able to enhance anticancer activity against lymph node metastases. The lymphatic pharmacokinetics and antitumor activity of PEGylated polylysine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service