WH0065220M1
Monoclonal Anti-NADK antibody produced in mouse
clone 5F4, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-FLJ13052, Anti-NAD kinase, Anti-RP1283E3.6, Anti-dJ283E3.1
About This Item
Recommended Products
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
5F4, monoclonal
form
buffered aqueous solution
species reactivity
rat, human, mouse
technique(s)
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG2aκ
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... NADK(65220)
General description
Immunogen
Sequence
MEMEQEKMTMNKELSPDAAAYCCSACHGDETWSYNHPIRGRAKSRSLSASPALGSTKEFRRTRSLHGPCPVTTFGPKACVLQNPQTIMHIQDPASQRLTWNKSPKSVLVIKKMRDASLLQPFKELCTHLMEENMIVYVEKKVLEDPAIASDESFGAVKKKFCTFREDYDDISNQIDFIICLGGDGTLLYASSLFQGSVPPVMAFHLGSLGFLTPFSFENFQSQVTQVIEGNAAVVLRSRLKVRVVKELRGKKTAVHNGLGEKGSQAAGLDMDVGKQAMQYQVLNEVVIDRGPSSYLSNVDVYLDGHLITTVQGDGVIVSTPTGSTAYAAAAGASMIHPNVPAIMITPICPHSLSFRPIVVPAGVELKIMLSPEARNTAWVSFDGRKRQEIRHGDSISITTSCYPLPSICVRDPVSDWFESLAQCLHWNVRKKQAHFEEEEEEEEEG
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service