Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500599

Sigma-Aldrich

Anti-CDCA2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

FLJ25804, MGC129906, MGC129907, cell division cycle associated 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 112 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDCA2(157313)

General description

Cell division cycle associated 2 (CDCA2) is expressed in the nucleus. The gene encoding it is localized on human chromosome 8p21.2. Anti-CDCA2 Antibody detects endogenous levels of total CDCA2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CDCA2.

Immunogen Range: 511-560

Biochem/physiol Actions

Cell division cycle associated 2 (CDCA2) dephosphorylates histone H3 and modulates DNA damage responses. It has roles in nuclear envelope reformation and chromatin reorganization. The protein has been shown to be upregulated in neuroblastoma tumors and melanoma cell lines.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pauline Lagarde et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(5), 608-615 (2013-01-16)
Synovial sarcoma (SS) occurs in both children and adults, although metastatic events are much more common in adults. Whereas the importance of the t(X;18) translocation in SS oncogenesis is well established, the genetic basis of SS metastasis is still poorly
Paola Vagnarelli et al.
Nucleus (Austin, Tex.), 3(2), 138-142 (2012-05-05)
Coordination of late mitotic events is crucial for the maintenance of genome stability and for the control of gene expression after cell division. Reversible protein phosphorylation regulates this process by de-phosphorylation of mitotic phospho-proteins in a sequential and coordinated manner:
Paola Vagnarelli
Advances in experimental medicine and biology, 773, 401-414 (2014-02-25)
Nuclear structure and chromatin changes are very useful biomarkers in cancer diagnosis. Despite this, their biological significance and relevance to cancer progression are still not well understood. The identification of new proteins that link the nuclear envelope to chromatin organization
Fumihiko Uchida et al.
PloS one, 8(2), e56381-e56381 (2013-02-19)
Cell division cycle associated 2 (CDCA2) recruits protein phosphatase 1 to chromatin to antagonize activation of ataxia telangiectasia mutated (ATM)-dependent signal transduction. ATM kinase plays a critical role in the DNA damage response and its phosphorylation cascade to inhibit the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service