Skip to Content
Merck
All Photos(1)

Key Documents

Z757039

Kern USB interface for DBS moisture analyzer

pkg of 1 ea

Synonym(s):

moisture sensor, moisture tester

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111502
NACRES:
NB.71

packaging

pkg of 1 ea

manufacturer/tradename

Kern DBS-A02

Application

USB Data interface kit (cable, driver, software - Balance Connection)

The UBS interface supports bidirectional data transfer from a moisture meter to a computer. This data exchange is asynchronous using ASCII - Code.
UBS data is conducted to a virtual port.

The scope of delivery for the USB interface cable includes a CD containing software drivers for creating the required virtual ports on the computer (software ‘Balance Connection KERN SCD 4.0‘ for the import of data to a PC program).

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service