Key Documents
TEM-CF150-AU-TH
Sigma-Aldrich
Carbon Film Supported Gold Square Mesh
Thick, size 150 mesh, box of 50
About This Item
UNSPSC Code:
41111761
size:
150 mesh
packaging:
box of 50
packaging
box of 50
size
150 mesh
General description
A 10 nm thick film of pure carbon supported standard squre mesh grids. Gold grid, 150 mesh.
Choose from one of the most recent versions:
Certificates of Analysis (COA)