Skip to Content
Merck
All Photos(1)

Key Documents

QAMINESET

QuicTPD Amine Screening Set

new
Sign Into View Organizational & Contract Pricing


About This Item

storage temp.

2-8°C

Quality Level

General description

QTPD Amine set comprised of 36 partial protacs plated in duplicate with rich linker diversity, architecture and length targeting Cereblon(CRBN) and von Hippel Lindau( VHL) E3 ligases ending in a amine terminal.

Application

This set can be used for reacting with warheads/ligands for targets of interest with solvent exposed pendant carboxylic acid handles via peptide coupling reactions. Two distinct warheads can be reacted with this set to generate 72 distinct small molecule degraders that can be used for subsequent screening.

Features and Benefits

Representative set of partial protacs targeting both CRBN and VHL with typical PEG, hydrophobic and rigid linkers that can be used for construction of PROTACs. Customers can make screen upto 24 reaction conditions with this set and empty glass shell vials provided with the pre-plated set. Glass shell vials allow for exploration of reaction conditions tolerant to different, reagents, solvents and temperature. Miniaturized synthesis allows for reduction in solvent, reagent, water and energy usage making this a sustainable and faster route to PROTAC discovery.

Other Notes

QuicTPD Amine Screening Set

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service