Skip to Content
Merck
Pentane-1-sulfonic acid sodium salt for ion pair chromatography, LiChropur™

Pentane-1-sulfonic acid sodium salt

Pentane-1-sulfonic acid sodium salt for ion pair chromatography, LiChropur™
Synonym(s):
Pentane-1-sulfonic acid sodium salt
Empirical Formula (Hill Notation):
C5H11NaO3S
CAS Number:
Molecular Weight:
174.19

Products

Brand
Formula Weight
Product Line
Purity
Quality Segment
Technique