Skip to Content
Merck
All Photos(1)

Key Documents

6.18508

Supelco

Pentane-1-sulfonic acid sodium salt

for ion pair chromatography, LiChropur

Synonym(s):

Pentane-1-sulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NaO3S
CAS Number:
Molecular Weight:
174.19
MDL number:
UNSPSC Code:
12161902
grade:
reagent grade

grade

reagent grade

product line

LiChropur

SMILES string

[Na+].[S](=O)(=O)([O-])CCCCC

InChI

1S/C5H12O3S.Na/c1-2-3-4-5-9(6,7)8;/h2-5H2,1H3,(H,6,7,8);/q;+1/p-1

InChI key

ROBLTDOHDSGGDT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service