Z331880
BRAND® Dispensette® III bottle-top dispenser, digital, Easy calibration
0.5-5 mL, with SafetyPrime
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121500
Recommended Products
feature
with SafetyPrime
manufacturer/tradename
BRAND 4700331 EU
BrandTech 4701331 US
measuring range
0.5-5 mL
Looking for similar products? Visit Product Comparison Guide
General description
Dispensette III (red color-code): for dispensing aggressive reagents, including concentrated acids such as H3PO4, H2SO4, bases like NaOH, KOH, saline solutions as well as many organic solvents - directly from a supply bottle.
The parts of the Dispensette Organic which are in contact with medium are made of: Borosilicate glass, ceramic, tantalum, ETFE, FEP & PFA. The discharge tube safety screw cap is PP.
Operating limits:
- Easy to dismantle for cleaning, with replaceable filling valves
- Autoclavable at 121°C
- Digital display: Especially easy to read and dispensing volume can be set accurately and reproducibly (mechanical counter)
- Easy Calibration: Innovative technique for adjustment in seconds without tools
- SafetyPrime™ recirculation valve reduces reagent waste and the safety discharge system reduces the risk of inadvertent dispensing and splashes
- To allow the bottle label to face the user, the valve block can be rotated 360°
- A telescopic filling tube adjusts easily to different size bottles
The parts of the Dispensette Organic which are in contact with medium are made of: Borosilicate glass, ceramic, tantalum, ETFE, FEP & PFA. The discharge tube safety screw cap is PP.
Operating limits:
- Vapor pressure max. 500 mbar
- Viscosity max. 500 mm2/s
- Temperature max. 40°C
- Density max. 2.2 g/cm3
- Accuracy: ±0.5%, precision ±0.1%
Features and Benefits
- Simple: Wide range of applications for organic solvents, acids, alkalis and saline solutions.
- Easy: Minimum operating forces, maximum comfort.
- Efficient: Reliable in challenging continuous operation and with aggressive media.
Linkage
See the Compatibility Chart to choose the correct bottle-top dispenser.
Other Notes
BR4700331 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
SafetyPrime is a trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service