Z328030
BRAND® Transferpette® pipette, digital, adjustable single channel
volume 100-1000 μL, accuracy: 0.6%, precision: 0.2%
All Photos(1)
About This Item
Recommended Products
feature
electronic: no
recommended tips 1000 μL & 1250 μL
* 50-1000 μL
single-channel
subdivision 1 μL
manufacturer/tradename
BRAND 704180
volume measuring range
100-1000 μL, accuracy: 0.6%, precision: 0.2%
volume
1000 μL
General description
The Transferpette pipette is designed to the shape of the human hand for maximum comfort. The side mounted pipetting key reduces hand fatigue, especially when performing serial pipetting operations. The textures housing of the hand grip ensures a firm grip and is UV resistant.
Changing volume is easy without snagging gloves. Indentations prevent inadvertent volume changes. Thanks to the Easy Calibration Technique calibration and adjustments according to ISO 9000 and GLP are done within seconds.
The slim shaft (autoclavable at 121°C) makes it possible to pipette in the narrowest vessels without removing the tip ejector. The taper is designed to fit PLASTIBRAND and most other leading manufacturers tips.
* Filter tips
Changing volume is easy without snagging gloves. Indentations prevent inadvertent volume changes. Thanks to the Easy Calibration Technique calibration and adjustments according to ISO 9000 and GLP are done within seconds.
The slim shaft (autoclavable at 121°C) makes it possible to pipette in the narrowest vessels without removing the tip ejector. The taper is designed to fit PLASTIBRAND and most other leading manufacturers tips.
* Filter tips
Other Notes
BR704180 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)