Skip to Content
Merck
All Photos(2)

Key Documents

SRP5099

Sigma-Aldrich

WNK1 (181-507), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

HSAN2, HSN2, KDP, KIAA0344, MGC163339, MGC163341, PRKWNK1, PSK, p65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

25-33 nmol/min·mg

mol wt

~67 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... WNK1(65125)

General description

WNK1 is a member of the WNK subfamily of serine/threonine protein kinases that is a key regulator of blood pressure by controlling the transport of sodium and chloride ions. Mutations in WNK1 have been associated with pseudohypoaldosteronism type II and hereditary sensory neuropathy type II. WNK1 is a regulator of blood pressure and deficiency of this protein in mice lowers the blood pressure. WNK1 can regulate the Ca(2+) sensing and the subsequent Ca(2+)-dependent interactions mediated by synaptotagmin C2 domains and WNK1 exhibits additive CFTR inhibition.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byung-Hoon Lee et al.
Molecular cell, 15(5), 741-751 (2004-09-08)
WNK (with no lysine [K]) protein kinases were named for their unique active site organization. Mutations in WNK1 and WNK4 cause a familial form of hypertension by undefined mechanisms. Here, we report that WNK1 selectively binds to and phosphorylates synaptotagmin
Brian P Zambrowicz et al.
Proceedings of the National Academy of Sciences of the United States of America, 100(24), 14109-14114 (2003-11-12)
The availability of both the mouse and human genome sequences allows for the systematic discovery of human gene function through the use of the mouse as a model system. To accelerate the genetic determination of gene function, we have developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service