GN188261
Greiner centrifuge tubes
15 mL, 17x120 mm, blue cap, conical bottom, clear tube
Synonym(s):
centrifuge tubes, culture tubes, plastic centrifuge tube, plastic culture tube, Greiner centrifuge tubes
About This Item
Recommended Products
material
polypropylene tube
sterility
sterile
packaging
pack of 500 ea
manufacturer/tradename
Greiner 188261
availability
available only in China
color
colorless tube
Looking for similar products? Visit Product Comparison Guide
General description
High quality, crystal-clear polypropylene centrifuge tubes. Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.
- Greiner centrifuge tubes are made of chemically resistant, high quality, crystal-clear polypropylene.
- Ideal for storing chemical and biological samples.
- Blue graduations facilitate precise volume measurements.
- White writing area for sample identification.
- Centrifuge tubes can be safely centrifuged to approximately 3,200 × g (50 mL size) and 3,500 × g (15 mL size) depending on the specific gravity of the solution being centrifuged and the type of rotor.
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic, non-cytotoxic.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service