AXYPCR384LC480CNF
PCR microplate for Roche® 480 LightCycler®
384 well, no sealing film, clear
Synonym(s):
384 multiwell plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
material:
clear
clear polypropylene wells
clear polypropylene wells
size:
384 wells
Recommended Products
material
clear
clear polypropylene wells
sterility
non-sterile
packaging
case of 50 ea
manufacturer/tradename
Axygen Inc
Corning
technique(s)
PCR: suitable
size
384 wells
working volume
5-20 mL
suitability
suitable for PCR
Looking for similar products? Visit Product Comparison Guide
Legal Information
LightCycler is a registered trademark of Roche
Roche is a registered trademark of Roche Molecular Systems, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service