Skip to Content
Merck
All Photos(1)

Key Documents

AV41669

Sigma-Aldrich

Anti-C2 (AB2) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CO2, Anti-Complement component 2, Anti-DKFZp779M0311

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

83 kDa

species reactivity

dog, rat, human, pig, mouse, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... C2(717)

Immunogen

Synthetic peptide directed towards the middle region of human C2

Biochem/physiol Actions

Component C2 is part of the classical pathway of complement system. Activated C1 cleaves C2 into C2a and C2b. C2a leads to activation of C3. Deficiency of C2 has been reported to associated with certain autoimmune diseases.Component C2 is part of the classical pathway of complement system. Activated C1 cleaves C2 into C2a and C2b. C2a leads to activation of C3. Deficiency of C2 has been reported to associated with certain autoimmune diseases.

Sequence

Synthetic peptide located within the following region: INQKRNDYLDIYAIGVGKLDVDWRELNELGSKKDGERHAFILQDTKALHQ

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service