Skip to Content
Merck
All Photos(1)

Key Documents

AV32347

Sigma-Aldrich

Anti-TLE2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ESG, Anti-ESG2, Anti-FLJ41188, Anti-GRG2, Anti-Transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

80 kDa

species reactivity

horse, rat, human, bovine, dog, pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLE2(7089)

General description

TLE-2 is the human homolog of Drosophila E(sp1) that interacts with the replication and transcription activator (RTA) protein. Studies have reported that TLE2 can block RTA-mediated transactivation and replication.
Rabbit Anti-TLE2 antibody recognizes human, mouse, and rat TLE2

Immunogen

Synthetic peptide directed towards the N terminal region of human TLE2

Application

Rabbit Anti-TLE2 antibody can be used for western blot applications at a concentration of 1μg/ml.

Biochem/physiol Actions

TLE2 belongs to the WD repeat Groucho/TLE family. It contains 6 WD repeats. TLE2 is a transcriptional corepressor that binds to a number of transcription factors. It inhibits the transcriptional activation mediated by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES.

Sequence

Synthetic peptide located within the following region: VEEERPSGPGGGGKQRADEKEPSGPYESDEDKSDYNLVVDEDQPSEPPSP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhiheng He et al.
Journal of virology, 84(4), 2047-2062 (2009-11-27)
Replication and transcription activator (RTA) encoded by open reading frame 50 (ORF50) of Kaposi's sarcoma-associated herpesvirus (KSHV) is essential and sufficient to initiate lytic reactivation. RTA activates its target genes through direct binding with high affinity to its responsive elements

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service