Skip to Content
Merck
All Photos(2)

Key Documents

68594

Supelco

Maslinic acid

analytical standard

Synonym(s):

2α,3β-Dihydroxyolean-12-en-28-oic acid, 2α-Hydroxyoleanolic acid, Crategolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O4
CAS Number:
Molecular Weight:
472.70
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(O)=O

InChI

1S/C30H48O4/c1-25(2)12-14-30(24(33)34)15-13-28(6)18(19(30)16-25)8-9-22-27(5)17-20(31)23(32)26(3,4)21(27)10-11-29(22,28)7/h8,19-23,31-32H,9-17H2,1-7H3,(H,33,34)/t19-,20+,21-,22+,23-,27-,28+,29+,30-/m0/s1

InChI key

MDZKJHQSJHYOHJ-LLICELPBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maslinic acid belongs to the class of triterpenic acids, a group of phytochemicals present in a large variety of plants. It exhibits a broad-spectrum of biological properties such as cardiovascular, antihyperlipidemic, antioxidant effects, hepatoprotective effects, antitumor activity. It is also involved in enhancing the cellular immune system.

Application

Maslinic acid may be used as an analytical reference standard for the quantification of the analyte in the leaves of Ziziphus species and fruits using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Predominant triterpenoid found in olives. Anti-proliferative. Promising chemopreventive agent.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service