Skip to Content
Merck
All Photos(1)

Key Documents

6.18226

Supelco

Paraffin liquid

light

Synonym(s):

Paraffin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12191502

form

liquid

viscosity

11 mm2/s(40 °C) (104 °F)

mp

<-15 °C (< 5 °F)

density

0.83 -0.86 g/cm3

Looking for similar products? Visit Product Comparison Guide

Application


  • Simple colloidal synthesis of single-crystal Sb-Se-S nanotubes with composition dependent band-gap energy in the near-infrared.: This study discusses a method involving the use of liquid paraffin in synthesizing single-crystal nanotubes, highlighting the impact of varying compositions on the band-gap energies in the near-infrared spectrum (Deng et al., 2009).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

419.0 °F - closed cup

Flash Point(C)

215 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service