Skip to Content
Merck
All Photos(1)

Key Documents

6.17857

Supelco

Silver nitrate solution

0.02 mol/l (0.02N)

Synonym(s):

Silver nitrate solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302

form

solution

SMILES string

[Ag]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Antimicrobial Activity of Silver Nanoparticles Synthesized Using Ocimum tenuiflorum and Ocimum gratissimum Herbal Formulations.: This paper demonstrates the utilization of silver nitrate solution for synthesizing silver nanoparticles with potent antimicrobial activities using herbal extracts. The study underscores the interdisciplinary approach in nanotechnology and herbal medicine, showcasing the application of silver nitrate solution in developing novel antimicrobial agents (Varghese et al., 2024).

  • Evaluation of Biogenic Silver Nanoparticles Synthesized from Vegetable Waste.: Highlighting the eco-friendly use of silver nitrate solution, this research focuses on the synthesis of biogenic silver nanoparticles from vegetable waste. The study offers insights into sustainable practices in nanoparticle synthesis, emphasizing the environmental and economic benefits of using silver nitrate solutions (Kiani et al., 2023).

  • Nanosphere Lithography-Enabled Hybrid Ag-Cu Surface-Enhanced Raman Spectroscopy Substrates with Enhanced Absorption of Excitation Light.: This research utilizes silver nitrate solution in the fabrication of hybrid Ag-Cu substrates for surface-enhanced Raman spectroscopy, improving the detection capabilities for chemical analysis. The application of silver nitrate solution in this context enhances the substrate′s light absorption properties, demonstrating its critical role in spectroscopic technologies (Wu et al., 2023).

  • Construction of silver-coated high translucent zirconia implanting abutment material and its property of antibacterial.: This article discusses the use of silver nitrate solution for creating silver-coated zirconia materials with enhanced antibacterial properties. The study highlights the importance of silver nitrate solution in dental applications, contributing to the development of advanced materials that prevent bacterial infections (Peng et al., 2023).

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service