Skip to Content
Merck
All Photos(1)

Key Documents

6.07590

Supelco

Resorcinol recryst.

Synonym(s):

Resorcinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OH)2
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352104

SMILES string

Oc1cc(ccc1)O

InChI

1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H

InChI key

GHMLBKRAJCXXBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service