Skip to Content
Merck
All Photos(1)

Key Documents

6.07553

Supelco

Extran® MA 02 neutral (for cleaning laboratory glassware & instruments)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161902

product line

Extran®

form

liquid

solubility

water: soluble

density

1.057 -1.067 g/cm3

application(s)

cleaning products

Legal Information

EXTRAN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service