Skip to Content
Merck
All Photos(1)

Key Documents

6.06076

Supelco

Methyl red (C.I. 13020)

pH indicator

Synonym(s):

Methyl Red, 2-(4-Dimethylaminophenylazo)benzoic acid, 4-Dimethylaminoazobenzene-2′-carboxylic acid, Acid Red 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4CO2H
CAS Number:
Molecular Weight:
269.30
Colour Index Number:
13020
Beilstein:
750102
MDL number:
UNSPSC Code:
12352107

Product Name

Methyl red indicator, C.I.No. 13020 S.No. 250

mp

179-182 °C (lit.)

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccccc2C(O)=O

InChI

1S/C15H15N3O2/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20/h3-10H,1-2H3,(H,19,20)/b17-16+

InChI key

CEQFOVLGLXCDCX-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service