524545
Phosphatase, Alkaline, E. coli
Synonym(s):
Phosphatase, Alkaline, E. coli
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
MDL number:
UNSPSC Code:
12352202
Recommended Products
form
liquid
Quality Level
specific activity
≥500 units/mL
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
pI
4.5
foreign activity
Non-specific endonuclease, RNase, none detected
shipped in
wet ice
storage temp.
−20°C
General description
Native alkaline phosphatase from E. coli. A dimeric zinc- and magnesium-containing protein. A non-specific phosphomonoesterase with a pI of 7.5.
Warning
Toxicity: Standard Handling (A)
Unit Definition
One unit is defined as the amount of enzyme that will hydrolyze 1 µmol of pNPP per min at 25°C in Tris-HCl, pH 8.0.
Physical form
In 50 mM Tris-HCl, 50% glycerol, pH 7.5.
Other Notes
Reid, T.W. and Willson, I.B. 1971. Enzymes4, 383.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service