Skip to Content
Merck
All Photos(1)

Key Documents

506101

Sigma-Aldrich

p21-Activated Kinase Inhibitor, PAK18

The p21-Activated Kinase Inhibitor, PAK18 controls the biological activity of p21-Activated Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

p21-Activated Kinase Inhibitor, PAK18, H₂N-RKKRRQRRR~G~PPVIAPRPEHTKSVYTRS-CO₂H, PAK Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C146H254N58O37
Molecular Weight:
3413.95
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A PAK (p21-activated kinase) inhibitor peptide that is composed of the cell permeant TAT peptide sequence and an 18-mer Pro-rich PIX-interacting motif of PAK that disrupts PIX-PAK interaction, reduces cellular PAK phosphorylation, and induces neurodegenerative morphology in hippocampal neurons in vitro. Intracerebroventricular administration of PAK18 in mice resulted in Alzheimer-like disease phenotype in vivo. PIX-interacting motif sequence has also been shown to block ras-dependent tumor growth and PAK activation in NIH-3T3 cells. The inactive control peptide, PAK18-R192A, is also available (Cat. No. 506102).

Biochem/physiol Actions

Cell permeable: no
Primary Target
PAK (p21-activated kinase)
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg~Gly~Pro-Pro-Val-Ile-Ala-Pro-Arg-Pro-Glu-His-Thr-Lys-Ser-Val-Tyr-Thr-Arg-Ser-OH

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service