Skip to Content
Merck
All Photos(1)

Key Documents

1.09258

Supelco

Karl Fischer′s solution pyridine-free single solution

Synonym(s):

Pyridine-Free KF Reagent

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352401

form

solution

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Corr. 1B - STOT RE 1 Oral - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus, Thyroid

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

98.6 °F

Flash Point(C)

37 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service