Key Documents
TEM-71152
Sigma-Aldrich
Grid Storage Box
capacity, 50 (wells), pack of 12 ea
Synonym(s):
GRID STORAGE BOX 50-CAP CLEAR COVER R13A2
About This Item
UNSPSC Code:
41111761
feature
capacity 50 (wells)
packaging
pack of 12 ea
General description
Grid Storage Box, 50 CapacityStorage for 50 grids in deep diamond-shaped wells. All wells are identified. The base is resistant to organics and reactions can be carried out on grid-mounted samples in the wells. Complete with grid recording card.
Choose from one of the most recent versions:
Certificates of Analysis (COA)