GF66009023
Nickel-chromium alloy, Ni80Cr20
100mm tube, 1.3mm outside diameter, 0.25mm wall thickness, 0.8mm inside diameter
Synonym(s):
NI057130
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
11101711
NACRES:
NA.23
form
tubes
manufacturer/tradename
Goodfellow 660-090-23
L × wall thickness
100 mm × 0.25 mm
O.D. × I.D.
1.3 mm × 0.8 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service