SAAR191165
Potassium permanganate 0.1N solution
Synonym(s):
Potassium permanganate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
glass bottle of 4 × 2.5 L (SAAR191165LC)
storage temp.
2-30°C
SMILES string
[K+].[O-][Mn](=O)(=O)=O
InChI
1S/K.Mn.4O/q+1;;;;;-1
InChI key
VZJVWSHVAAUDKD-UHFFFAOYSA-N
Application
For laboratory use only
Suitable for heterogeneous oxidations.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service