WH0055607M1
Monoclonal Anti-PPP1R9A antibody produced in mouse
clone 6A10, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-FLJ20068, Anti-KIAA1222, Anti-NRB1, Anti-NRBI, Anti-NeurabinI, Anti-protein phosphatase 1, regulatory (inhibitor) subunit 9A
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6A10, monoclonal
form
buffered aqueous solution
species reactivity
human, rat, mouse
technique(s)
indirect ELISA: suitable
western blot: 1-5 μg/mL
isotype
IgG1κ
GenBank accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PPP1R9A(55607)
Immunogen
Sequence
PCSTAQTSTRSPCMPFSWFNDSRKGSYSFRNLPAPTSSLQPSPETLISDKKGSKNFTFNDDFSPSSTSSADLSGLGAEPKTPGLSQSLALSSDESLDMI
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service