R0906
RSK3, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
HU-2, MAPKAPK1 C, RPS6KA2, SK6K-alpha, p90rsk3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
111-151 nmol/min·mg
mol wt
~112 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... RPS6KA2(6196)
Biochem/physiol Actions
RSK3 is a member of the RSK (ribosomal S6 kinase) family that encodes a 733-amino-acid protein with a unique N-terminal region containing a putative nuclear localization signal. RSK3 mRNA is widely expressed and is activated by growth factors, serum and phorbol ester. Upon stimulation, RSK3 translocates to the cell nucleus and phosphorylates nuclear proteins. RSK3 can bind to ERK1/2 and this association increases the duration of RSK3 activation.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Y Zhao et al.
Molecular and cellular biology, 15(8), 4353-4363 (1995-08-01)
A novel pp90rsk Ser/Thr kinase (referred to as RSK3) was cloned from a human cDNA library. The RSK3 cDNA encodes a predicted 733-amino-acid protein with a unique N-terminal region containing a putative nuclear localization signal. RSK3 mRNA was widely expressed
Philippe P Roux et al.
Molecular and cellular biology, 23(14), 4796-4804 (2003-07-02)
Stimulation of the Ras/extracellular signal-regulated kinase (ERK) pathway can modulate cell growth, proliferation, survival, and motility. The p90 ribosomal S6 kinases (RSKs) comprise a family of serine/threonine kinases that lie at the terminus of the ERK pathway. Efficient RSK activation
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service