Skip to Content
Merck
All Photos(5)

Key Documents

PLA0307

Sigma-Aldrich

Goat anti-Sp1 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

specificity protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:1,000-1:5,000
immunoprecipitation (IP): 2- 10 μg/mg
western blot: 1:2,000- 1:10,000

accession no.

NP_612482.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... Sp1(6667)

Immunogen

The epitope recognized by PLA0307 maps to a region between residue 735 to 785 of human Sp1 Transcription Factor using the numbering given in entry NP_612482.2 (GeneID 6667).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Specificity protein 1 (Sp1) is the original member of the Sp1-like/KLF family of zinc-finger transcription factors that binds GC-rich DNA elements to regulate transcription of genes involved in a variety of cellular functions. Sp-1 interacts with coactivators such as p300/CBP, TAFII130, and CRSP.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepti Malik et al.
Molecular cancer, 13, 175-175 (2014-07-20)
microRNAs (miRNAs) play both oncogenic and oncostatic roles in leukemia. However, the molecular details underlying miRNA-mediated regulation of their target genes in pediatric B- and T-cell acute lymphoblastic leukemias (ALLs) remain unclear. The present study investigated the relationship between miR-2909
Carolina Vizcaíno et al.
PloS one, 9(8), e104687-e104687 (2014-08-12)
Ovarian cancer has a poor prognosis due to intrinsic or acquired resistance to some cytotoxic drugs, raising the interest in new DNA-binding agents such as mithramycin analogues as potential chemotherapeutic agents in gynecological cancer. Using a genome-wide approach, we have
Luigi Formisano et al.
Journal of neuroscience research, 93(1), 167-177 (2014-08-06)
Polychlorinated biphenyls (PCBs) cause a wide spectrum of toxic effects in the brain through undefined mechanisms. Exposure to the PCB mixture Aroclor-1254 (A1254) increases the repressor element-1 silencing transcription factor (REST) expression, leading to neuronal death. This study sought to
D Koyama et al.
Leukemia, 28(6), 1216-1226 (2013-12-05)
The Notch signaling pathway has been recognized as a key factor for the pathogenesis of T-cell acute lymphoblastic leukemia (T-ALL), because of the high incidence of activating mutations of Notch1. Notch inhibition could serve as a new treatment strategy for
Ting Wei et al.
Journal of biomedical science, 21, 87-87 (2014-09-04)
Lipid accumulation is the primary evidence of non-alcoholic fatty liver disease (NAFLD). Ginkgo biloba extract (GBE) and its flavonoid ingredients (quercetin, kaempferol, and isorhamnetin) could lessen the lipid accumulation associated with up-regulation of the rate-limiting enzyme, carnitine palmitoyltransferase 1A (CPT1A)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service