Skip to Content
Merck
All Photos(1)

Key Documents

AV38544

Sigma-Aldrich

Anti-PITX3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Paired-like homeodomain transcription factor 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

32 kDa

species reactivity

mouse, horse, human, rat, pig, rabbit

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PITX3(5309)

Immunogen

Synthetic peptide directed towards the N terminal region of human PITX3

Biochem/physiol Actions

PITX3 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. Mutations of PITX3 have been associated with anterior segment mesenchymal dysgenesis (ASMD) and congenital cataracts. PITX3 is involved in lens formation during eye development.This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. Mutations of this gene have been associated with anterior segment mesenchymal dysgenesis (ASMD) and congenital cataracts. This protein is involved in lens formation during eye development.

Sequence

Synthetic peptide located within the following region: ARSPALSLSDAGTPHPQLPEHGCKGQEHSDSEKASASLPGGSPEDGSLKK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Soeki et al.
Heart and vessels, 29(5), 653-658 (2013-08-28)
Increasing evidence indicates that inflammation contributes to the pathogenesis of atrial fibrillation (AF). Pentraxin 3 (PTX3) is produced abundantly in local inflammatory lesions while C-reactive protein (CRP) is produced mainly in the liver. In this study, we investigated whether a
Anna Pertek et al.
Molecular biotechnology, 56(8), 697-713 (2014-03-29)
Mammalian cells can be reprogrammed into induced pluripotent stem cells (iPSCs), a valuable tool for in vitro disease modeling and regenerative medicine. These applications demand for iPSCs devoid of reprogramming factor transgenes, but current procedures for the derivation of transgene-free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service