Skip to Content
Merck
All Photos(1)

Key Documents

03-0250

Sigma-Aldrich

Barium chloride dihydrate

SAJ first grade, ≥98.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCl2 · 2H2O
CAS Number:
Molecular Weight:
244.26
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.5%

form

crystalline

availability

available only in Japan

pH

5.0-8.0 (25 °C, 50 g/L)

SMILES string

O.O.Cl[Ba]Cl

InChI

1S/Ba.2ClH.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

PWHCIQQGOQTFAE-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anne R Crecelius et al.
The Journal of physiology, 590(21), 5349-5359 (2012-07-11)
Circulating ATP possesses unique vasomotor properties in humans and has been hypothesized to play a role in vascular control under a variety of physiological conditions. However, the primary downstream signalling mechanisms underlying ATP-mediated vasodilatation remain unclear. The purpose of the
Rajani M George et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(46), 18549-18554 (2013-10-31)
The adaptor protein Numb has been implicated in the switch between cell proliferation and differentiation made by satellite cells during muscle repair. Using two genetic approaches to ablate Numb, we determined that, in its absence, muscle regeneration in response to
Laura A Merriam et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(10), 4614-4622 (2013-03-08)
After G-protein-coupled receptor activation and signaling at the plasma membrane, the receptor complex is often rapidly internalized via endocytic vesicles for trafficking into various intracellular compartments and pathways. The formation of signaling endosomes is recognized as a mechanism that produces
Fan Hu et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 19(12), 1051-1058 (2012-08-15)
Danshen (Salviae Miltiorrhizae Radix) and Gegen (Puerariae Lobatae Radix) have been widely used in treating cardiovascular diseases for thousands of years in China. The present study was carried out to evaluate the effects of a Danshen and Gegen decoction (DG)
Chung-Lin Wang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 30(3), 687-701 (2012-08-03)
Mitoxanthrone (MX) is an anthracenedione antineoplastic agent. Whether this drug and other related compounds have any effects on ion currents in osteoclasts remains largely unclear. In this study, the effects of MX and other related compounds on inwardly rectifying K(+)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service