Skip to Content
Merck
All Photos(1)

Key Documents

8.41514

Sigma-Aldrich

Diethyl acetylenedicarboxylate

for synthesis

Synonym(s):

Diethyl acetylenedicarboxylate, 2-Butynedioic acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10O4
CAS Number:
Molecular Weight:
170.16
MDL number:
UNSPSC Code:
12352108
EC Index Number:
212-095-8

Quality Level

Assay

≥97.0% (GC)

form

liquid

bp

108-110 °C/14.7 hPa

mp

1.5 °C

transition temp

flash point 94 °C

density

1.067 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H10O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-4H2,1-2H3

InChI key

STRNXFOUBFLVIN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.063 - 1.073
Identity (IR): passes test

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F - closed cup

Flash Point(C)

94 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service