Key Documents
TEM-FF150CUUL
Sigma-Aldrich
Ultrathin Formvar Supported Copper Square Mesh
size 150 mesh, box of 50
Synonym(s):
FORMVAR SQUARE MESH - CU 150 MESH UL 50/BX
About This Item
UNSPSC Code:
41111761
packaging
box of 50
size
150 mesh
General description
A 5-6 nm ultrathin formvar film supported standard copper square mesh grids. Copper grid, 150 mesh. All of our film is laid on the shiny side of the grid.
Choose from one of the most recent versions:
Certificates of Analysis (COA)