Skip to Content
Merck
All Photos(2)

Key Documents

V800127

Sigma-Aldrich

Cobalt(II) sulfate heptahydrate

LR, ≥98%

Synonym(s):

Cobaltous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CoSO4 · 7H2O
CAS Number:
Molecular Weight:
281.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥98%
grade:
LR
form:
crystalline

grade

LR

product line

Vetec

Assay

≥98%

form

crystalline

pH

4 (20 °C, 100 g/L)

density

2.03 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.01%

cation traces

Fe: ≤0.01%

SMILES string

O.[Co++].[O-]S([O-])(=O)=O

InChI

1S/Co.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

BGORGFZEVHFAQU-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B Inhalation - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P J Duerksen-Hughes et al.
Environmental health perspectives, 107(10), 805-812 (1999-09-30)
In vivo carcinogenicity testing is an expensive and time-consuming process, and as a result, only a relatively small fraction of new and existing chemicals has been tested in this manner. Therefore, the development and validation of alternative approaches is desirable.
Ali M Elshafei et al.
Journal of basic microbiology, 45(5), 335-343 (2005-09-28)
Cell-free extracts of nitrate-grown Penicillium politans NRC-510 catalyzes the hydrolytic deamination of cytidine to uridine. Uridine was chromatographically identified in cell-free extracts. The enzyme exhibited optimum pH and temperature activities at 6.5 and 80 degrees C respectively. Thermal stability experiments
Hong Wang et al.
ACS nano, 7(1), 614-626 (2012-12-12)
Electronic and optical properties of single-walled carbon nanotubes (SWCNTs) correlate with their chiral structures. Many applications need chirally pure SWCNTs that current synthesis methods cannot produce. Here, we show a sulfate-promoted CoSO(4)/SiO(2) catalyst, which selectively grows large-diameter (9,8) nanotubes at
Y J Huang et al.
Chemosphere, 40(4), 347-349 (2000-02-09)
A rapid and complete destruction of organics in the supercritical water oxidation (SCWO) of the Co-exchanged resin was found experimentally. Due to an extremely low solubility of CoSO4 salt formed and separated effectively in the SCWO process, a minimal release
Carolyn F Moyer et al.
Toxicologic pathology, 30(4), 427-434 (2002-08-22)
Epidemiological studies suggest an association between ambient particulate matter and cardiopulmonary diseases in humans. The mechanisms underlying these health effects are poorly understood. To better understand the potential relationship between particulate-matter-induced inflammation and vascular disease, a 2-phase retrospective study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service