M1941
Myelin Basic Protein Kinase 2 from bovine kidney
>90% (SDS-PAGE)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
bovine kidney
Quality Level
Assay
>90% (SDS-PAGE)
form
solution
specific activity
≥1000 units/mg protein
mol wt
40 kDa
shipped in
dry ice
storage temp.
−70°C
Unit Definition
One unit will incorporate one nanomole of phosphate into myelin basic protein at pH 7.0 at 30 °C
Physical form
Solution at 1 μg/mL in 50 mM Tris-HCl, pH 7.0, 1 mM benzamidine, 1 mM EDTA, 0.1 mM PMSF, 14 mM β-mercaptoethanol, and 50% glycerol.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service