Recommended Products
Product Name
WIL2.NS.6TG, 93031001
biological source
human spleen
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblast
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human Caucasian B lymphocyte
Cell Line Description
A non Ig-secreting, HAT sensitive, 6-thioguanine resistant, human B lymphocyte. Derived from the parent line WIL2.NS (Sigma Catalogue number. 90112121).
Application
Immortalising antigen-stimulated human B-lymphocytes by somatic hybridisation
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 5x1,000-5x10,000 cells/ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service