Skip to Content
Merck
All Photos(2)

Key Documents

C5149

Sigma-Aldrich

Calcium Ionophore A23187 mixed calcium magnesium salt

Approximate 1:1 molar ratio Ca:Mg. Actual content given on label.

Synonym(s):

Calcimycin, Calcium Ionophore mixed calcium magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37N3O6
Molecular Weight:
523.62
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

99% (TLC)

form

powder

solubility

chloroform/methanol: 10 mg/mL, clear, faintly yellow

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Mg++].[Ca++].CNc1ccc2oc(CC3OC4(CCC3C)OC(C(C)CC4C)C(C)C(=O)c5ccc[nH]5)nc2c1C(O)=O

InChI

1S/C29H37N3O6.Ca.Mg/c1-15-10-11-29(17(3)13-16(2)27(38-29)18(4)26(33)20-7-6-12-31-20)37-22(15)14-23-32-25-21(36-23)9-8-19(30-5)24(25)28(34)35;;/h6-9,12,15-18,22,27,30-31H,10-11,13-14H2,1-5H3,(H,34,35);;/q;2*+2

InChI key

HKVXGTQLKZNXPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Calcium Ionophore A23187 mixed calcium magnesium salt has been used as a calcium ionophore to increase the intracellular levels of calcium.
Ionophore highly selective for Ca2+. Potentiates responses to NMDA but not to quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability of the acrosome of the brush-tailed possum (Trichosurus vulpecula) and tammar wallaby (Macropus eugenii) in vitro and after exposure to conditions and agents known to cause capacitation or acrosome reaction of eutherian spermatozoa.
Mate KE and Rodger JC
Journal of Reproduction and Fertility, 91, 41-48 (1991)
Method of transfection affects the cAMP-mediated induction of the RIIbeta subunit of protein kinase A in Sertoli cells: inhibition of response by increase in intracellullar calcium.
Gronning LM, et al.
European Journal of Endocrinology, 141, 75-82 (1999)
Fatima El-Assaad et al.
PLoS pathogens, 10(3), e1003839-e1003839 (2014-03-22)
In patients with cerebral malaria (CM), higher levels of cell-specific microparticles (MP) correlate with the presence of neurological symptoms. MP are submicron plasma membrane-derived vesicles that express antigens of their cell of origin and phosphatidylserine (PS) on their surface, facilitating
Reiya Taniguchi et al.
Nature communications, 6, 8545-8545 (2015-10-16)
In vertebrates, the iron exporter ferroportin releases Fe(2+) from cells into plasma, thereby maintaining iron homeostasis. The transport activity of ferroportin is suppressed by the peptide hormone hepcidin, which exhibits upregulated expression in chronic inflammation, causing iron-restrictive anaemia. However, due
Arthur Liesz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(2), 583-598 (2015-01-16)
Acute brain lesions induce profound alterations of the peripheral immune response comprising the opposing phenomena of early immune activation and subsequent immunosuppression. The mechanisms underlying this brain-immune signaling are largely unknown. We used animal models for experimental brain ischemia as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service