Skip to Content
Merck
All Photos(1)

Key Documents

36018

Supelco

Fehling′s reagent I for sugars

reag. Ph. Eur., solution I: copper(II) sulfate

Synonym(s):

Copper(II) sulfate solution, Cupric sulfate solution acc. to Fehling, Cupric sulfate standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
reag. Ph. Eur.

Quality Level

concentration

17.6 g/L±0.1 g/L Cu

density

1.038-1.060 g/mL at 20 °C

suitability

in accordance for application

application(s)

food and beverages
general analytical
pharmaceutical (small molecule)

SMILES string

[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The principle behind Fehling′ test is basically based on the reducing property of monosaccharide and disaccharides, which in turn depends on the presence of free keto or an aldehyde group. This test is performed by adding a few drops of sugar solution to a mixture of Fehling′s A solution (i.e., CuSO4) and Fehling′s B solution (i.e., KOH and Na-K tartarate). Fehling′s solution gets reduced by aldose sugars to insoluble yellow or red colored cuprous oxide. Monosaccharides like glucose, galactose and fructose and disaccharides like lactose and maltose resond freely to Fehling′s test as they possess a free keto or an aldehyde group, whereas sucrose fails to answer the test as it does not possess a free aldehyde or an keto group. Hence, Fehling′s test is also used to distinguish reducing discaccharides like maltose from non-reducing sugars like sucrose.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elementary Practical Organic Chemistry: Quantitative Organic Analysis Part 3.
Voguel A.
Science, 743-744 null
Lab Manual in Biochemistry: Immunology and Biotechnology.
Tata N.
Biochemistry, 23-23 (2007)
Xu-Chen Cao et al.
PloS one, 8(2), e56735-e56735 (2013-03-08)
The aquaporin (AQP) family consists of a number of small integral membrane proteins that transport water and glycerol. AQPs are critical for trans-epithelial fluid transport. Recent reports demonstrated that AQPs, particularly AQP1 and AQP5, are expressed in high grade tumor
M Seder-Colomina et al.
Ecotoxicology (London, England), 22(1), 199-205 (2012-11-10)
Microbial mats are coastal ecosystems that consist mainly of cyanobacteria, primary producers in these habitats that play an important role in stabilising delta sediments. However, these ecosystems are subject to various kinds of pollution, including metal contamination, placing their survival
Antonella Amore et al.
FEMS microbiology letters, 337(2), 155-163 (2012-10-12)
In silico analyses of several laccase promoter sequences have shown the presence of many different responsive elements differentially distributed along the promoter sequences. Analysis of Pleurotus ostreatus laccase promoter poxa1b extending around 1400-bp upstream of the start codon showed the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service