AB4344
Anti-LYAR Antibody (Ly1 Antibody reactive clone)
from rabbit, purified by affinity chromatography
Synonym(s):
Cell growth-regulating nucleolar protein, Protein expressed in male leptotene and zygotene spermatocytes 264, MLZ-264
About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
mouse
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
mouse ... Lyar(17089)
General description
Immunogen
Application
Immunoglobulins & Immunology
Pluripotent & Early Differentiation
Quality
Western Blot Analysis: A 1:1,000 dilution of this antibody detected LYAR in 10 µg of mouse embryonic stem cell lysate.
Target description
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service