8.10158
Chlorodiphenylphosphine
for synthesis
Synonym(s):
Chlorodiphenylphosphine, Diphenylchlorophosphine
About This Item
Recommended Products
vapor pressure
1.3 hPa ( 20 °C)
Quality Level
Assay
≥97.0% (GC)
form
liquid
autoignition temp.
320 °C
potency
316 mg/kg LD50, oral (Rat)
bp
320 °C/1013 hPa
mp
14-16 °C
transition temp
flash point 138 °C
density
1.20 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/C12H10ClP/c13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H
InChI key
XGRJZXREYAXTGV-UHFFFAOYSA-N
Analysis Note
Density (d 20 °C/ 4 °C): 1.192 - 1.196
Identity (IR): passes test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
Supplementary Hazards
Storage Class Code
8A - Combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
280.4 °F
Flash Point(C)
138 °C
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service