Skip to Content
Merck
All Photos(1)

Key Documents

SML1462

Sigma-Aldrich

Practolol

≥95% (HPLC)

Synonym(s):

(±)-Practolol, 1-(4-Acetamidophenoxy)-3-isopropylamino-2-propanol, 4′-[2-Hydroxy-3-(isopropylamino)propoxy]acetanilidex, AY 21011, Dalzic, ICI 50172, N-[4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O3
CAS Number:
Molecular Weight:
266.34
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC(NC1=CC=C(OCC(O)CNC(C)C)C=C1)=O

InChI

1S/C14H22N2O3/c1-10(2)15-8-13(18)9-19-14-6-4-12(5-7-14)16-11(3)17/h4-7,10,13,15,18H,8-9H2,1-3H3,(H,16,17)

InChI key

DURULFYMVIFBIR-UHFFFAOYSA-N

Biochem/physiol Actions

Practolol is a potent and selective β-adrenoceptor antagonist. Practolol is a beta blocker that was discontinued to due to adverse reactions (keratoconjunctivitis sicca, conjunctival scarring, fibrosis, metaplasia, and shrinkage).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Qiao Cheng et al.
Molecules (Basel, Switzerland), 24(4) (2019-03-03)
This paper presents an application of high performance liquid chromatography coupled with quadrupole orbitrap high-resolution mass spectrometry (HPLC-Q-Orbitrap HRMS) for the analysis of 27 β-blockers and metabolites in milk powder. Homogenized milk power samples were extracted by acetonitrile and purified
Guohua Huang et al.
Combinatorial chemistry & high throughput screening, 19(2), 121-128 (2015-11-11)
It is crucial to identify the molecular targets of a compound during the course of the new drug discovery and drug development. Due to the complexity of biological systems, finding drug targets by biological experiments is very tedious and expensive.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service