Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408184

Sigma-Aldrich

Anti-CHRDL1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CHL, NRLN1, VOPT, dA141H5.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~51.2 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHRDL1(91851)

General description

This gene encodes an antagonist of bone morphogenetic protein 4. The encoded protein may play a role in topographic retinotectal projection and in the regulation of retinal angiogenesis in response to hypoxia. Alternatively spliced transcript variants encoding different isoforms have been described. (provided by RefSeq)

Immunogen

CHRDL1 (ENSP00000361115, 1 a.a. ~ 450 a.a) full-length human protein.

Sequence
MGGMKYIFSLLFFLLLEGGKTEQVKHSETYCMFQDKKYRVGERWHPYLEPYGLVYCVNCICSENGNVLCSRVRCPNVHCLSPVHIPHLCCPRCPDSLPPVNNKVTSKSCEYNGTTYQHGELFVAEGLFQNRQPNQCTQCSCSEGNVYCGLKTCPKLTCAFPVSVPDSCCRVCRGDGELSWEHSDGDIFRQPANREARHSYHRSHYDPPPSRQAGGLSRFPGARSHRGALMDSQQASGTIVQIVINNKHKHGQVCVSNGKTYSHGESWHPNLRAFGIVECVLCTCNVTKQECKKIHCPNRYPCKYPQKIDGKCCKVCPGKKAKELPGQSFDNKGYFCGEETMPVYESVFMEDGETTRKIALETERPPQVEVHVWTIRKGILQHFHIEKISKRMFEELPHFKLVTRTTLSQWKIFTEGEAQISQMCSSRVCRTELEDLVKVLYLERSEKGHC

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jelena Katic et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(44), 14606-14623 (2014-10-31)
The cell adhesion molecule close homolog of L1 (CHL1) plays important functional roles in the developing and adult nervous system. In search of the binding partners that mediate the diverse and sometimes opposing functions of CHL1, the extracellular matrix-associated proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service