Skip to Content
Merck
All Photos(1)

Key Documents

L5412

Sigma-Aldrich

Anti-Lymphotoxin β Receptor antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-LT-βR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
conjugate:
unconjugated
application:
FACS
WB
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
flow cytometry: 2.5 μg using 106 cells
western blot: 0.1-0.2 μg/mL

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

flow cytometry: 2.5 μg using 106 cells
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... LTBR(4055)

General description

The antibody will recognize recombinant human LT-βR. Applications include in immunoblotting, flow cytometry, and agonist activity.

Immunogen

recombinant human lymphotoxin β receptor extracellular domain expressed in NS0 cells.

Biochem/physiol Actions

Lymphotoxins (LT) alpha and beta are homologs of tumor necrosis factor. These cytokines play important role in the development and function of immune system, immune response and autoimmunity. The signaling mediated by LTβ receptor mediates autoimmune reactions and the development of secondary lymphoid organs.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline containing carbohydrates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hugh McDevitt et al.
Arthritis research, 4 Suppl 3, S141-S152 (2002-07-12)
Tumor necrosis factor (TNF)-alpha and lymphotoxin (LT) alpha/beta play multiple roles in the development and function of the immune system. This article focuses on three important aspects of the effects of these cytokines on the immune response and on autoimmunity.
Role of lymphotoxin in experimental models of infectious diseases: potential benefits and risks of a therapeutic inhibition of the lymphotoxin-beta receptor pathway.
Thomas W Spahn et al.
Infection and immunity, 73(11), 7077-7088 (2005-10-22)
H Körner et al.
Immunology and cell biology, 74(5), 465-472 (1996-10-01)
Tumour necrosis factor (TNF) is a highly potent, proinflammatory cytokine with broad-ranging functions from the regulation of endothelial cell adhesion molecules to facilitate entry of leucocytes into tissues, to direct induction of cellular cytotoxicity. This diversity of function potentially attributable
Yu Mikami et al.
PloS one, 9(12), e114791-e114791 (2014-12-17)
Asthma-related mortality has been decreasing due to inhaled corticosteroid use, but severe asthma remains a major clinical problem. One characteristic of severe asthma is resistance to steroid therapy, which is related to neutrophilic inflammation. Recently, the tumor necrosis factor superfamily

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service