EP0030509234
Eppendorf® Deepwell plates, Protein LoBind, 96 wells
capacity 1000 μL, pkg of 80 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
capacity:
1000 μL
size:
96 wells
packaging
pkg of 80 ea
manufacturer/tradename
Eppendorf® 0030509234
technique(s)
PCR: suitable
capacity
1000 μL
size
96 wells
Looking for similar products? Visit Product Comparison Guide
General description
Maximize your protein sample recovery with Eppendorf Protein LoBind Plates. Eppendorf LoBind Plates are equipped with a special two-component polymer mix that creates a hydrophilic surface to ensure optimal sample recovery rates. These plates are specifically designed for protein research or sensitive testing methods in proteomics, where they enable excellent results due to less sample binding to the plate surface.
Application
- Preparation or storage of protein, peptide or antibody samples
- Enzymatic assays – the hydrophilic surface reduces denaturation when interacting with the vessel wall
- Avoiding sample loss when storing virus stock solutions
- Storage of cell suspensions
Features and Benefits
- Eppendorf LoBind material ensures outstanding sample recovery for better test results
- Without surface coating (e.g. silicone) to minimize the risk of sample interference
- Batch-certified purity grade PCR clean: free from human DNA, DNase, RNase and PCR inhibitors
- Available as reaction vessel, microplate and deepwell plate for easy scaling
- Unique high-contrast OptiTrack matrix: up to 30% faster sample detection and fewer pipetting errors
- RecoverMax well design: Optimized well geometry for minimized residual or dead volume and excellent mixing properties
- Raised corrugated edges and a smooth surface ensure reliable sealing of the plates
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service