Skip to Content
Merck
All Photos(1)

Key Documents

HSCRMAG32PMX14BK

Millipore

MILLIPLEX® Human Soluble Cytokine Receptor Panel, HSCRMAG-32K- Space Saver (Bulk) Packaging

Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

sensitivity: 4-208 pg/mL
(Overnight Incubation minDC)

standard curve range: 12.2-50,000 pg/mL
(sIL-4R, sIL-6R, sRAGE, sTNFRI, sTNFRII)

standard curve range: 122.1-500,000 pg/mL
(sEGFR, sIL-1RII, sVEGF-R1, sVEGF-R2, sVEGF-R3)

standard curve range: 24.4-100,000 pg/mL
(sCD30, sgp130, sIL-1RI, sIL-2Rα)

inter-assay cv: <15%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

compatibility

configured for Premixed

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cytokine receptors constitute an integral part of cytokine biology. Like cytokines, cytokine receptors are involved in normal physiological and pathological processes of almost all disease states. Soluble cytokine receptors naturally arise from genes encoding membrane- bound receptors or are direct derivatives of the receptors themselves. The discovery that soluble cytokine receptors are involved in regulating excessive inflammatory responses and modulating immune events has stimulated significant research interest in their potential role as immunotherapeutic agents. Many of these soluble cytokine receptors can inhibit the binding and biological activity of their cytokine ligands, making them very specific cytokine antagonists.

MILLIPLEX® Human Soluble Cytokine Receptor Panel is to be used for the simultaneous quantification of 14 soluble cytokine receptors in human serum, plasma and cell / tissue culture supernatant samples. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cytokines/Chemokines

Specificity

Cross Reactivty
There is no detectable cross-reactivity within the panel.
UPDATE >>
Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • Analytes: sCD30, sEGFR, sGP130, sIL-1RI, sIL-1RII, sIL-2Rα, sIL-4R, sIL-6R, sRAGE, sTNFRI, sTNFRII, sVEGFR1, sVEGFR2, sVEGFR3
  • Recommended Sample type: Serum, plasma or tissue/cell lysate and culture supernatant
  • Recommended Sample dilution: 1:5 dilution for plasma or serum or neat cell culture supernatant
  • Assay Run Time: Overnight
  • Research Category: Inflammation & Immunology

Storage and Stability

Recommended storage for kit components is 2 - 8°C.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: Please see kit protocol for individual assay sensitivities.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service