Recommended Products
composition
(refer to Biolab Culture Media Catalogue for composition)
packaging
poly bottle of 12 × 500 g (HG00C104.500)
pH
5.0±0.2 (25 °C)
storage temp.
15-25°C
Application
Preparation Note
Do not Autoclave or Overheat
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service