Skip to Content
Merck
All Photos(1)

Key Documents

8.21139

Sigma-Aldrich

Chlorotriethylsilane

for synthesis

Synonym(s):

Chlorotriethylsilane, Triethylchlorosilane, Triethylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3SiCl
CAS Number:
Molecular Weight:
150.72
MDL number:
UNSPSC Code:
12352103
EC Index Number:
213-615-6

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

280 °C

bp

144-145 °C/1013 hPa

transition temp

flash point 30 °C

density

0.90 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H15ClSi/c1-4-8(7,5-2)6-3/h4-6H2,1-3H3

InChI key

DCFKHNIGBAHNSS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.897 - 0.900
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F

Flash Point(C)

30 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service